Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Plastic drum, 20 L
Catalog # 20910.443
Health
Toxic
Corrosive
Supplier:   VWR Chemicals
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®
Catalog # 20910.443
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Plastic drum, 20 L

CAS Number:  50-00-0
Price
1.730,00 kr
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1,088 g/cm³ (20 °C)
  • Flash Pt:66 °C (closed cup)
  • Storage Temperature:Stue temperatur
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
  •  :stabiliseret med methanol 10 %
Specifications

Specification Test Results

  • Assay:
    35 - 38 %
  • Density (20/4):
    1.083 - 1.093
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Methanol:
    8 - 12 %

Specifications

  • Pakning:
    Plastic drum
  • Pakn:
    20 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.