Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Plastic bottle, 1 L
Katalog # 20910.294
Sundhed
Giftig
Ætsende
Leverandør:   VWR Chemicals
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®
Katalog # 20910.294
Leverandør:  VWR Chemicals
CAS nummer:  50-00-0
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®
Formaldehyd 36% in aqueous solution stabilised, GPR RECTAPUR®

Plastic bottle, 1 L

CAS nummer:  50-00-0
Price
209,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS nummer:50-00-0
  • Formel:CH₂O
  • MDL nummer:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Tæthed:1,088 g/cm³ (20 °C)
  • Flash Pt:66 °C (closed cup)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-001-8
  • FN:2209
  • ADR:8,III
  •  :stabiliseret med methanol 10 %
Specifikationer

Specifikationstestresultater

  • Assay:
    35 - 38 %
  • Density (20/4):
    1.083 - 1.093
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Methanol:
    8 - 12 %

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 L

Specifikationer