ISA opløsninger
Nitrate interference suppressor solution (NISS) for nitrate ion selective electrode, for more accurate and reproducible ISE measurements, 475 ml bottle, Ion selective electrode reagents, Bottle, 475 mL
Katalog # 662-0812
Leverandør: Thermo Orion
Gå til:
Product Details & Documents
Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
- Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
- Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
- Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
- Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences
Concentration : 10 ppm
For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
Documents
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:475 mL
- Beskrivelse:Nitrate interference suppressor solution (NISS) for nitrate ion selective electrode, for more accurate and reproducible ISE measurements, 475 ml bottle
- Model:Ion selective electrode reagents
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



