Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-2-Butanol ≥99% analytical reagent

(±)-2-Butanol ≥99% analytical reagent

Glass bottle, 1 L
Katalog # 20812.295
Brandfarlig
Irriterende
Leverandør:   VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
Katalog # 20812.295
Leverandør:  VWR Chemicals
CAS nummer:  78-92-2
(±)-2-Butanol ≥99% analytical reagent
(±)-2-Butanol ≥99% analytical reagent

Glass bottle, 1 L

CAS nummer:  78-92-2
Price
416,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
  • CAS nummer:78-92-2
  • Formel:H₃CCH₂CH(OH)CH₃
  • MDL nummer:MFCD00004569
  • Molecular Weight:74.12 g/mol
  • Merck Index:13,01540
  • Kogende Pt:94 °C (1013 hPa)
  • Smeltende Pt:–115 °C
  • Tæthed:0,8534 g/cm³ (20 °C)
  • Flash Pt:23 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-158-5
  • FN:1120
  • ADR:3,III
Specifikationer

Specifikationstestresultater

  • Assay (on anhydrous substance):
    Min. 99.0 %
  • Acidity:
    Max. 0.0005 meq/g
  • Boiling point:
    98.5 - 100.5 °C
  • Density (20/4):
    0.805 - 0.809
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 0.2 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.