Orders LinkContactUsLinkComponent
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
Nitrogen nitrate reagent set, 0 - 0,50 mg/l NO₃- N, Cadmium reduction, DR3000, DR2000, 25 ml, PP, 100, 100 Tests
Katalog # HACH2245100
Leverandør:   Hach
Nitrogen-nitrate reagent sets, LR
Katalog # HACH2245100
Leverandør:  Hach
CAS nummer:  
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
Nitrogen nitrate reagent set, 0 - 0,50 mg/l NO₃- N, Cadmium reduction, DR3000, DR2000, 25 ml, PP, 100, 100 Tests
CAS nummer:  
Price
1.780,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
For determination of nitrogen-nitrate by the cadmium reduction method.
Specifikationer
Specifikationer
  • Kemisk metode:
    Cadmium reduction
  • Pakn:
    100 Tests
  • Beskrivelse:
    Nitrogen nitrate reagent set
  • Prøvestørrelse:
    25 ml
  • Model:
    PP
  • Til enhed:
    DR3000, DR2000
  • Test pr. sæt:
    100
  • Område:
    0 - 0,50 mg/l NO₃- N