Orders LinkContactUsLinkComponent
Powder pillows, PathoScreen Medium Presence/Absence
PathoScreen medium MPN pillows, 20 ml samples, 50
  HACH2610796
 :   Hach
Powder pillows, PathoScreen Medium Presence/Absence
  HACH2610796
 :  Hach
 :  
Powder pillows, PathoScreen Medium Presence/Absence
Powder pillows, PathoScreen Medium Presence/Absence
PathoScreen medium MPN pillows, 20 ml samples, 50
 :  
707,00 kr
1 * 50 Items
These are used to detect waterborne pathogens associated with fecal contamination of water sources.
  • Simple presence-absence method eliminates need for an incubator in tropical and semi-tropical climates
  • Complete Solutions for testing drinking water
  • Pakn:
    50
  • Beskrivelse:
    PathoScreen medium MPN pillows, 20 ml samples