Powder pillows, PathoScreen Medium Presence/Absence
PathoScreen medium presence / absence powder pillows, 100 ml samples, 50
HACH2610696
: Hach
These are used to detect waterborne pathogens associated with fecal contamination of water sources.
- Simple presence-absence method eliminates need for an incubator in tropical and semi-tropical climates
- Complete Solutions for testing drinking water
- Pakn:50
- Beskrivelse:PathoScreen medium presence / absence powder pillows, 100 ml samples