Orders LinkContactUsLinkComponent
Chlorophosphonazo indicator solution for hardness determination
Chlorophosphonazo indicator solution for hardness determination
Chlorophosphonazo indicator solution pillows, 100 per pack, 100
Catalog # HACH2589599
Supplier:   Hach
Chlorophosphonazo indicator solution for hardness determination
Catalog # HACH2589599
Supplier:  Hach
CAS Number:  
Chlorophosphonazo indicator solution for hardness determination
Chlorophosphonazo indicator solution for hardness determination
Chlorophosphonazo indicator solution pillows, 100 per pack, 100
CAS Number:  
Price
3.670,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
For the determination of ultra low range hardness by chlorophosphonazo method. Ideal for use with Pour-Thru cell. Requires CDTA reagent.
Specifications
Specifications
  • Pakn:
    100
  • Beskrivelse:
    Chlorophosphonazo indicator solution pillows, 100 per pack
Specifications