Chlorophosphonazo indicator solution for hardness determination
Chlorophosphonazo indicator solution pillows, 100 per pack, 100
Catalog # HACH2589599
Supplier: Hach
Chlorophosphonazo indicator solution for hardness determination
Catalog # HACH2589599
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
For the determination of ultra low range hardness by chlorophosphonazo method. Ideal for use with Pour-Thru cell. Requires CDTA reagent.
Specifications
Specifications
- Pakn:100
- Beskrivelse:Chlorophosphonazo indicator solution pillows, 100 per pack