Orders LinkContactUsLinkComponent
EC medium MPN tubes
EC/MUG without durham tubes, -, 15
  HACH2471515
 :   Hach
EC medium MPN tubes
  HACH2471515
 :  Hach
 :  
EC medium MPN tubes
EC medium MPN tubes
EC/MUG without durham tubes, -, 15
 :  
604,00 kr
1 * 15 Items
EC medium is used to detect coliform bacteria using the Most Probable Number (MPN) method.
  • Save time and labour costs with ready to use MPN tubes, with minimal training requirements
  • Pakn:
    15
  • Standard:
    -
  • Beskrivelse:
    EC/MUG without durham tubes