Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

EDTA ≥99% analytical reagent

Plastic bottle for solids, 1 kg
Katalog # 20301.290
Irriterende
Leverandør:   VWR Chemicals
EDTA ≥99% analytical reagent
Katalog # 20301.290
Leverandør:  VWR Chemicals
CAS nummer:  60-00-4
EDTA ≥99% analytical reagent
EDTA ≥99% analytical reagent

Plastic bottle for solids, 1 kg

CAS nummer:  60-00-4
Price
2.090,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
  • CAS nummer:60-00-4
  • Formel:C₁₀H₁₆N₂O₈
  • MDL nummer:MFCD00003541
  • Molecular Weight:292.25 g/mol
  • Merck Index:13,03546
  • Kogende Pt:614 °C (1013 hPa)
  • Smeltende Pt:220 °C
  • Tæthed:0,86 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-449-4
  • NÅ:01-2119486399-18
Specifikationer

Specifikationstestresultater

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.2 %
  • Water:
    Max. 0.2 %
  • Cl (Chloride):
    Max. 0.05 %
  • SO4 (Sulphate):
    Max. 0.05 %
  • Fe (Iron):
    Max. 10 ppm

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    1 kg

Specifikationer