Borsyre ≥98%, TECHNICAL
Plastic bucket, 5 kg
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- CAS nummer:10043-35-3
- Formel:B(OH)₃
- MDL nummer:MFCD00011337
- Molecular Weight:61.83 g/mol
- Merck Index:13,01326
- Kogende Pt:185 °C (1013 hPa)
- Smeltende Pt:160 °C
- Tæthed:1,44 g/cm³ (25 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:233-139-2
- NÅ:01-2119486683-25
Documents
Certifikater
Søg efter partinummer1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 5L Plastic bucket UN Z (ZPACK13247005)
Emballageoplysninger
28041714.pdf P20_VWRC20177.368_SPEC_EN.pdf spec_VWRC20177.290 spec_VWRC20177.368
Produktspecifikation
Specifikationer
Specifikationstestresultater
- Assay:Min. 98 %
Specifikationer
- Pakning:Plastic bucket
- Pakn:5 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



