Orders LinkContactUsLinkComponent
EDTA solutions
EDTA solutions
EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration, Self-contained dropping bottle, 1 L
Katalog # HACH20553
Leverandør:   Hach
EDTA solutions
Katalog # HACH20553
Leverandør:  Hach
CAS nummer:  
EDTA solutions
EDTA solutions
EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration, Self-contained dropping bottle, 1 L
CAS nummer:  
Price
314,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Documents
Specifikationer
Specifikationer
  • Pakning:
    Self-contained dropping bottle
  • Pakn:
    1 L
  • Beskrivelse:
    EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration