Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Molybdate reagent

Molybdate reagent

Molybdate reagent for phosphate determination, 1 l, 1 L
Katalog # HACH223653
Leverandør:   Hach
Molybdate reagent
Katalog # HACH223653
Leverandør:  Hach
CAS nummer:  
Molybdate reagent
Molybdate reagent

Molybdate reagent for phosphate determination, 1 l, 1 L

CAS nummer:  
Price
448,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Determination of high range phosphate (phosphorus) by the amino acid method.

Documents

Specifikationer

Specifikationer

  • Pakn:
    1 L
  • Beskrivelse:
    Molybdate reagent for phosphate determination, 1 l

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.