Orders LinkContactUsLinkComponent
Chlorine standard solution
Chlorine standard solution
Chlorine standard solution, 10 ml Voluette ampoules, 50 - 75 mg/l as Cl₂, 16
Katalog # 705-0060
Leverandør:   Hach
Chlorine standard solution
Katalog # 705-0060
Leverandør:  Hach
CAS nummer:  
Chlorine standard solution
Chlorine standard solution
Chlorine standard solution, 10 ml Voluette ampoules, 50 - 75 mg/l as Cl₂, 16
CAS nummer:  
Price
1.030,00 kr
Log ind for tilgængelighed
1 * 16 Items
Product Details & Documents
For accuracy checks in chlorine determinations. Lot specific concentration displayed on label.
  • Simple accuracy checks
  • Minimise waste
  • Helps ensure reliable result
Specifikationer
Specifikationer
  • Koncentration:
    50 - 75 mg/l as Cl₂
  • Pakn:
    16
  • Beskrivelse:
    Chlorine standard solution, 10 ml Voluette ampoules