Orders LinkContactUsLinkComponent
Phosphate standard solutions
Phosphate standard solutions
Phosphate standard solution, 946 ml (NIST), 10 mg/l as PO4, 946 mL
Catalog # HACH1420416
Supplier:   Hach
Phosphate standard solutions
Catalog # HACH1420416
Supplier:  Hach
CAS Number:  
Phosphate standard solutions
Phosphate standard solutions
Phosphate standard solution, 946 ml (NIST), 10 mg/l as PO4, 946 mL
CAS Number:  
Price
514,00 kr
Log in for availability
1 * 946 mL
Product Details & Documents
Standard solution for accuracy checks in phosphate (phosphorus) determinations.
Specifications
Specifications
  • Koncentration:
    10 mg/l as PO<sub>4</sub>
  • Pakn:
    946 mL
  • Beskrivelse:
    Phosphate standard solution, 946 ml (NIST)