EC medium MPN tubes
EC medium MPN tubes, -, 15
Catalog # HACH1410415
Supplier: Hach
Jump to:
Product Details & Documents
EC medium is used to detect coliform bacteria using the Most Probable Number (MPN) method.
- Save time and labour costs with ready to use MPN tubes, with minimal training requirements
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pakn:15
- Standard:-
- Beskrivelse:EC medium MPN tubes