Orders LinkContactUsLinkComponent
EC medium MPN tubes
EC medium MPN tubes
EC medium MPN tubes, -, 15
Catalog # HACH1410415
Supplier:   Hach
EC medium MPN tubes
Catalog # HACH1410415
Supplier:  Hach
CAS Number:  
EC medium MPN tubes
EC medium MPN tubes
EC medium MPN tubes, -, 15
CAS Number:  
Price
529,00 kr
Log in for availability
1 * 15 Items
Product Details & Documents
EC medium is used to detect coliform bacteria using the Most Probable Number (MPN) method.
  • Save time and labour costs with ready to use MPN tubes, with minimal training requirements
Documents
Specifications
Specifications
  • Pakn:
    15
  • Standard:
    -
  • Beskrivelse:
    EC medium MPN tubes
Specifications