Orders LinkContactUsLinkComponent
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
Nitrogen nitrate reagent set, 0,1 - 0,50 mg/l NO₃- N, Cadmium reduction, All except DR3000, DR2010, DR2000, PC II, 10 ml, PP, 100, 100
Katalog # 705-0132
Leverandør:   Hach
Nitrogen-nitrate reagent sets, LR
Katalog # 705-0132
Leverandør:  Hach
CAS nummer:  
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
Nitrogen nitrate reagent set, 0,1 - 0,50 mg/l NO₃- N, Cadmium reduction, All except DR3000, DR2010, DR2000, PC II, 10 ml, PP, 100, 100
CAS nummer:  
Price
1.380,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
For determination of nitrogen-nitrate by the cadmium reduction method.
Specifikationer
Specifikationer
  • Kemisk metode:
    Cadmium reduction
  • Pakn:
    100
  • Beskrivelse:
    Nitrogen nitrate reagent set
  • Prøvestørrelse:
    10 ml
  • Model:
    PP
  • Til enhed:
    All except DR3000, DR2010, DR2000, PC II
  • Test pr. sæt:
    100
  • Område:
    0,1 - 0,50 mg/l NO₃- N