Orders LinkContactUsLinkComponent
Empty solid phase extraction cartridges, Bond Elut™
Patron, 1 ml
  551-0356
 :   VARIAN
Empty solid phase extraction cartridges, Bond Elut™
  551-0356
 :  VARIAN
Empty solid phase extraction cartridges, Bond Elut™
Empty solid phase extraction cartridges, Bond Elut™
Patron, 1 ml
1.050,00 kr
1 * 100 Items
Bond Elut empty SPE cartridges are made with high purity polypropylene for cleaner extracts.
  • Uniform batch-to-batch size for consistent performance
  • Cartridges are available from 1 to 60 ml
 : Frits has to be ordered separately
  • Kolonne form:
    Patron
  • Kapacitet:
    1 ml
  • Seq:
    01
Accessories
  •  
    Beskrivelse
  • 551-0354
    Adapter caps for 1, 3 and 6 ml Bond Elut™ cartridges
    1 * 15 Items - 1.030,00 kr
  •  
    Beskrivelse
  • 551-0355
    Luer stopcocks
    1 * 15 Items - 1.490,00 kr
  •  
    Beskrivelse
  • VARI12131035
    Adapter caps for3 ml Gilson ASPEC SPE systems
    1 * 1.000 Items - 8.400,00 kr
  •  
    Beskrivelse
  • VARI12131034
    Adapter caps for1ml Gilson ASPEC SPE systems
    1 * 1.000 Items - 7.720,00 kr