Empty solid phase extraction cartridges with two frits, Bond Elut™
Empty cartridge with two frits, Patron, PP, 12 ml
551-0364
: VARIAN
Clean PP reservoirs contain two PP frits pre-inserted, an ideal configuration for simple filtration.
- Pre-installed frits for ease-of-use
- Broad range of filtration operations for maximum flexibility
- Customisable packing for specific applications
- Kolonne form:Patron
- Kapacitet:12 ml
- Beskrivelse:Empty cartridge with two frits
- Seq:05
- Kolonnemateriale:PP