Orders LinkContactUsLinkComponent
Empty solid phase extraction cartridges with two frits, Bond Elut™
Empty cartridge with two frits, Patron, PP, 12 ml
  551-0364
 :   VARIAN
Empty solid phase extraction cartridges with two frits, Bond Elut™
  551-0364
 :  VARIAN
Empty solid phase extraction cartridges with two frits, Bond Elut™
Empty solid phase extraction cartridges with two frits, Bond Elut™
Empty cartridge with two frits, Patron, PP, 12 ml
2.300,00 kr
1 * 100 Items
Clean PP reservoirs contain two PP frits pre-inserted, an ideal configuration for simple filtration.
  • Pre-installed frits for ease-of-use
  • Broad range of filtration operations for maximum flexibility
  • Customisable packing for specific applications
  • Kolonne form:
    Patron
  • Kapacitet:
    12 ml
  • Beskrivelse:
    Empty cartridge with two frits
  • Seq:
    05
  • Kolonnemateriale:
    PP