Orders LinkContactUsLinkComponent
Skalpelblade
Nr. 6
  233-0032
Skalpelblade
  233-0032
Skalpelblade
Skalpelblade
Nr. 6
410,00 kr
1 * 100 Items
Kulstofstål.
  • Available in sterile individual foil packs or non sterile foil packs of five
  • Quick and easy to assemble
  • Model:
    Nr. 6
  • Seq:
    01