Orders LinkContactUsLinkComponent
Visual comparator test kits, iron
Iron test kit, colour disc model IR-18 B, 0,25 - 7 mg/l, 1,10-Phenanthroline, 100, 1 KIT
Katalog # HACH146401
Leverandør:   Hach
Visual comparator test kits, iron
Katalog # HACH146401
Leverandør:  Hach
CAS nummer:  
Visual comparator test kits, iron
Visual comparator test kits, iron
Iron test kit, colour disc model IR-18 B, 0,25 - 7 mg/l, 1,10-Phenanthroline, 100, 1 KIT
CAS nummer:  
Price
1.310,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifikationer
Specifikationer
  • Kemisk metode:
    1,10-Phenanthroline
  • Pakn:
    1 KIT
  • Beskrivelse:
    Iron test kit, colour disc model IR-18 B
  • Test pr. sæt:
    100
  • Område:
    0,25 - 7 mg/l