[EN]GLASS DOOR 1 * 1 Items
Katalog # JOUALT89000020
Leverandør: JOUAN
[EN]GLASS DOOR 1 * 1 Items
Product Details & Documents
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
Leverandør: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
Leverandør: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
#: 1.00030.2500
Leverandør: Merck
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
Leverandør: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
Leverandør: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
#: 1.09439.1000
Leverandør: Merck



