Orders LinkContactUsLinkComponent
Multiparameter test kits, model HA-62
Multiparameter test kits, model HA-62
For hardness (as CaCO₃), iron and pH, HA-62, Hardness: 17,19 - 342,36 mg/l, Iron: 0,1 - 5 mg/l, pH: 6,5 - 8,5, 1 KIT
Katalog # HACH183700
Leverandør:   Hach
Multiparameter test kits, model HA-62
Katalog # HACH183700
Leverandør:  Hach
CAS nummer:  
Multiparameter test kits, model HA-62
Multiparameter test kits, model HA-62
For hardness (as CaCO₃), iron and pH, HA-62, Hardness: 17,19 - 342,36 mg/l, Iron: 0,1 - 5 mg/l, pH: 6,5 - 8,5, 1 KIT
CAS nummer:  
Price
3.470,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
  • Popular tests in one rugged kit
  • Uses color disc and drop count titration methods
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    For hardness (as CaCO₃), iron and pH, HA-62
  • Område:
    Hardness: 17,19 - 342,36 mg/l, Iron: 0,1 - 5 mg/l, pH: 6,5 - 8,5
Specifikationer