Orders LinkContactUsLinkComponent
VWR® Drum Keys
VWR® Drum Keys
Drum key, metal, for seals and 2" and 3/4" closures on metal drums
Katalog # VWRFCLEUNIVMETAL
Leverandør:   VWR Collection
VWR® Drum Keys
Katalog # VWRFCLEUNIVMETAL
Leverandør:  VWR Collection
VWR® Drum Keys
VWR® Drum Keys
Drum key, metal, for seals and 2" and 3/4" closures on metal drums
Price
544,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
A selection of tools to assist in opening IBCs, 200 litre drums, plus bottles and other containers with DIN 32, 45, 54 and 80 caps.
Specifikationer
  • Beskrivelse:
    Drum key, metal, for seals and 2" and 3/4" closures on metal drums