Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Zinkchlorid ≥97%, TECHNICAL

Zinkchlorid ≥97%, TECHNICAL

Plastic bucket, 5 kg
Katalog # 29126.363
Irriterende
Ætsende
Miljø
Leverandør:   VWR Chemicals
Zinkchlorid ≥97%, TECHNICAL
Katalog # 29126.363
Leverandør:  VWR Chemicals
CAS nummer:  7646-85-7
Zinkchlorid ≥97%, TECHNICAL
Zinkchlorid ≥97%, TECHNICAL

Plastic bucket, 5 kg

CAS nummer:  7646-85-7
Price
1.620,00 kr
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents
Synonymer:  Zinc dichloride, Dichlorozinc, Zinc (II) chloride
  • CAS nummer:7646-85-7
  • Formel:ZnCl₂
  • MDL nummer:MFCD00011295
  • Molecular Weight:136.3 g/mol
  • Merck Index:13,10185
  • Kogende Pt:732 °C (1013 hPa)
  • Smeltende Pt:283 °C
  • Tæthed:2,91 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-592-0
  • FN:2331
  • ADR:8,III
Specifikationer

Specifikationstestresultater

  • Assay:
    Min. 97 %
  • Fe (Iron):
    Max. 20 ppm
  • Pb (Lead):
    Max. 20 ppm

Specifikationer

  • Pakning:
    Plastic bucket
  • Pakn:
    5 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.