Orders LinkContactUsLinkComponent
Zinkchlorid ≥97%, TECHNICAL
Zinkchlorid ≥97%, TECHNICAL
Plastic bottle for solids, 1 kg
Katalog # 29126.294
Irriterende
Ætsende
Miljø
Leverandør:   VWR Chemicals
Zinkchlorid ≥97%, TECHNICAL
Katalog # 29126.294
Leverandør:  VWR Chemicals
CAS nummer:  7646-85-7
Zinkchlorid ≥97%, TECHNICAL
Zinkchlorid ≥97%, TECHNICAL
Plastic bottle for solids, 1 kg
CAS nummer:  7646-85-7
Price
438,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Zinc dichloride, Dichlorozinc, Zinc (II) chloride
  • CAS nummer:7646-85-7
  • Formel:ZnCl₂
  • MDL nummer:MFCD00011295
  • Molecular Weight:136.3 g/mol
  • Merck Index:13,10185
  • Kogende Pt:732 °C (1013 hPa)
  • Smeltende Pt:283 °C
  • Tæthed:2,91 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-592-0
  • FN:2331
  • ADR:8,III
Specifikationer
Specifikationstestresultater
  • Assay:
    Min. 97 %
  • Fe (Iron):
    Max. 20 ppm
  • Pb (Lead):
    Max. 20 ppm
Specifikationer
  • Pakning:
    Plastic bottle for solids
  • Pakn:
    1 kg
Specifikationer