Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Isooctan ≥98%, TECHNICAL

Isooctan ≥98%, TECHNICAL

Metal drum, 25 L
Katalog # 28775.461
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   VWR Chemicals
Isooctan ≥98%, TECHNICAL
Katalog # 28775.461
Leverandør:  VWR Chemicals
CAS nummer:  540-84-1
Isooctan ≥98%, TECHNICAL
Isooctan ≥98%, TECHNICAL

Metal drum, 25 L

CAS nummer:  540-84-1
Price
6.200,00 kr
Log ind for tilgængelighed
1 * 25 L
Product Details & Documents
Synonymer:  Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
  • CAS nummer:540-84-1
  • Formel:(CH₃)₂CHCH₂C(CH₃)₃
  • MDL nummer:MFCD00008943
  • Molecular Weight:114.23 g/mol
  • Merck Index:13,05212
  • Kogende Pt:99,2 °C (1013 hPa)
  • Smeltende Pt:–107 °C
  • Tæthed:0,692 g/cm³ (20 °C)
  • Flash Pt:–12 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-759-1
  • FN:1262
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Assay:
    Min. 98 %
  • Refractive index (20°C):
    1.391 - 1.393

Specifikationer

  • Pakning:
    Metal drum
  • Pakn:
    25 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.