Isooctan ≥98%, TECHNICAL
Metal can, 5 L
: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
- :540-84-1
- :(CH₃)₂CHCH₂C(CH₃)₃
- :MFCD00008943
- :114.23 g/mol
- :13,05212
- :99,2 °C (1013 hPa)
- :–107 °C
- :0,692 g/cm³ (20 °C)
- :–12 °C
- :Stue temperatur
- :208-759-1
- :1262
- :3,II
Documents
1L Glass bottle for liquids DIN 45 (ZPACK13415101) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Tin bottle for liquids UN Y (ZPACK12321007)
Packaging Information
P20_VWRC28775.290_SPEC_EN.pdf P20_VWRC28775.368_SPEC_EN.pdf spec_VWRC28775.290 spec_VWRC28775.290 spec_VWRC28775.368
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
- Assay:Min. 98 %
- Refractive index (20°C):1.391 - 1.393
- Pakning:Metal can
- Pakn:5 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



