BOD bottles, WHEATON®
BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 01-36, 60 ml
Katalog # WHEA227494-01
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specifikationer
- Kapacitet:60 ml
- Beskrivelse:BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 01-36