Natriumhydroxid ≥98%, pellets, GPR RECTAPUR®
Plastic bottle for solids, 5 kg
Leverandør: VWR Chemicals
Natriumhydroxid ≥98%, pellets, GPR RECTAPUR®
Katalog # 28245.367
Leverandør: VWR Chemicals
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Kogende Pt:1390 °C (1013 hPa)
- Smeltende Pt:323 °C
- Tæthed:2,13 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-185-5
- FN:1823
- ADR:8,II
- NÅ:01-2119457892-27
Documents
Certifikater
Søg efter partinummer1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Emballageoplysninger
P20_VWRC28245.265_SPEC_EN.pdf P20_VWRC28245.298_SPEC_EN.pdf P20_VWRC28245.367_SPEC_EN.pdf P20_VWRC28245.460_SPEC_EN.pdf spec_VWRC28245.265
Produktspecifikation
More
VWR Chemicals Big Packs and Bulks
Produktlitteratur
Specifikationer
Specifikationstestresultater
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 5 ppm
- Total N (Nitrogen):Max. 5 ppm
- CO3 (as Na2CO3):Max. 1.0 %
- Cl (Chloride):Max. 50 ppm
- PO4 (Phosphate):Max. 20 ppm
- SiO4 (as SiO2):Max. 100 ppm
- SO4 (Sulphate):Max. 50 ppm
- Al (Aluminium):Max. 10 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 0.1 %
- Pb (Lead):Max. 5 ppm
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:5 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



