Natriumhydroxid ≥98%, pellets, GPR RECTAPUR®
Plastic bottle for solids, 1 kg
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :14,08627
- :1390 °C (1013 hPa)
- :323 °C
- :2,13 g/cm³ (20 °C)
- :Stue temperatur
- :215-185-5
- :1823
- :8,II
- :01-2119457892-27
1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 5 ppm
- Total N (Nitrogen):Max. 5 ppm
- CO3 (as Na2CO3):Max. 1.0 %
- Cl (Chloride):Max. 50 ppm
- PO4 (Phosphate):Max. 20 ppm
- SiO4 (as SiO2):Max. 100 ppm
- SO4 (Sulphate):Max. 50 ppm
- Al (Aluminium):Max. 10 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 0.1 %
- Pb (Lead):Max. 5 ppm
- Pakning:Plastic bottle for solids
- Pakn:1 kg



