Natriumhydroxid ≥98%, pellets, GPR RECTAPUR®
Plastic bottle for solids, 500 g
Leverandør: VWR Chemicals
Natriumhydroxid ≥98%, pellets, GPR RECTAPUR®
Katalog # 28245.265
Leverandør: VWR Chemicals
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Kogende Pt:1390 °C (1013 hPa)
- Smeltende Pt:323 °C
- Tæthed:2,13 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-185-5
- FN:1823
- ADR:8,II
- NÅ:01-2119457892-27
Certifikater
Søg efter partinummer1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Specifikationer
Specifikationstestresultater
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 5 ppm
- Total N (Nitrogen):Max. 5 ppm
- CO3 (as Na2CO3):Max. 1.0 %
- Cl (Chloride):Max. 50 ppm
- PO4 (Phosphate):Max. 20 ppm
- SiO4 (as SiO2):Max. 100 ppm
- SO4 (Sulphate):Max. 50 ppm
- Al (Aluminium):Max. 10 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 0.1 %
- Pb (Lead):Max. 5 ppm
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:500 g



