Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®

Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®

Plastic drum, 20 L
Katalog # 28224.445
Ætsende
Leverandør:   VWR Chemicals
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®
Katalog # 28224.445
Leverandør:  VWR Chemicals
CAS nummer:  1310-73-2
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®

Plastic drum, 20 L

CAS nummer:  1310-73-2
Price
3.240,00 kr
Log ind for tilgængelighed
1 * 20 L
Product Details & Documents
Synonymer:  Caustic soda
  • CAS nummer:1310-73-2
  • Formel:NaOH
  • MDL nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Kogende Pt:119…135 °C (1013 hPa)
  • Smeltende Pt:9…10 °C
  • Tæthed:∼1,38 g/ml
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-185-5
  • FN:1824
  • ADR:8,II
Specifikationer

Specifikationstestresultater

  • Assay:
    34 - 36 %
  • Density (20/4):
    1.370 - 1.390
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm

Specifikationer

  • Pakning:
    Plastic drum
  • Pakn:
    20 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.