Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®

Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®

Plastic bottle, 5 L
Catalog # 28224.365
Corrosive
Supplier:   VWR Chemicals
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®
Catalog # 28224.365
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®

Plastic bottle, 5 L

CAS Number:  1310-73-2
Price
1.120,00 kr
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:∼1,38 g/ml
  • Storage Temperature:Stue temperatur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay:
    34 - 36 %
  • Density (20/4):
    1.370 - 1.390
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm

Specifications

  • Pakning:
    Plastic bottle
  • Pakn:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.