Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®
Plastic bottle, 1 L
Leverandør: VWR Chemicals
Natriumhydroxid 35% in aqueous solution, GPR RECTAPUR®
Katalog # 28224.296
Leverandør: VWR Chemicals
CAS nummer: 1310-73-2
Gå til:
Product Details & Documents
Synonymer: Caustic soda
- CAS nummer:1310-73-2
- Formel:NaOH
- MDL nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Kogende Pt:119…135 °C (1013 hPa)
- Smeltende Pt:9…10 °C
- Tæthed:∼1,38 g/ml
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-185-5
- FN:1824
- ADR:8,II
Documents
Certifikater
Søg efter partinummer1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Emballageoplysninger
Specifikationer
Specifikationstestresultater
- Assay:34 - 36 %
- Density (20/4):1.370 - 1.390
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 0.02 %
- Fe (Iron):Max. 20 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



