Natriumsulfat ≥98%, TECHNICAL
Plastic bottle for solids, 1 kg
: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
- :7757-82-6
- :Na₂SO₄
- :MFCD00003504
- :142.04 g/mol
- :12,08829
- :1429 °C (1013 hPa)
- :888 °C
- :2,7 g/cm³ (20 °C)
- :Stue temperatur
- :231-820-9
- Assay:Min. 98 %
- Pakning:Plastic bottle for solids
- Pakn:1 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



