Natriumchlorid ≥98%, TECHNICAL
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Saline, NaCl, Sea salt, Halite
- CAS Number:7647-14-5
- Formula:NaCl
- MDL Number:MFCD00003477
- Molecular Weight:58.44 g/mol
- Merck Index:14,08599
- Boiling Pt:1413 °C (1013 hPa)
- Melting Pt:801 °C
- Density:2,16 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:231-598-3
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
P20_VWRC27788.297_SPEC_EN.pdf P20_VWRC27788.366_SPEC_EN.pdf P20_VWRC27788.460_SPEC_EN.pdf spec_VWRC27788.297 spec_VWRC27788.297
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 98 %
- Appearance:Colourless crystals
Specifications
- Pakning:Plastic bottle for solids
- Pakn:1 kg