Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tetrachlorethylen ≥98%, TECHNICAL

Tetrachlorethylen ≥98%, TECHNICAL

Plastic bottle, fluorinated, 5 L
Catalog # 26217.367
Health
Environment
Supplier:   VWR Chemicals
Tetrachlorethylen ≥98%, TECHNICAL
Catalog # 26217.367
Supplier:  VWR Chemicals
CAS Number:  127-18-4
Tetrachlorethylen ≥98%, TECHNICAL
Tetrachlorethylen ≥98%, TECHNICAL

Plastic bottle, fluorinated, 5 L

CAS Number:  127-18-4
Price
1.120,00 kr
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  • CAS Number:127-18-4
  • Formula:C₂Cl₄
  • MDL Number:MFCD00000834
  • Molecular Weight:165.83 g/mol
  • Merck Index:12,09332
  • Boiling Pt:121,1 °C (1013 hPa)
  • Melting Pt:–22 °C
  • Density:1,63 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:204-825-9
  • UN:1897
  • ADR:6.1,III
Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Refractive index (20°C):
    1.503 - 1.505

Specifications

  • Pakning:
    Plastic bottle, fluorinated
  • Pakn:
    5 L

Specifications