Tetrachlorethylen ≥98%, TECHNICAL
Glass bottle, 2,5 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
- CAS nummer:127-18-4
- Formel:C₂Cl₄
- MDL nummer:MFCD00000834
- Molecular Weight:165.83 g/mol
- Merck Index:12,09332
- Kogende Pt:121,1 °C (1013 hPa)
- Smeltende Pt:–22 °C
- Tæthed:1,63 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-825-9
- FN:1897
- ADR:6.1,III
Documents
Certifikater
Søg efter partinummer1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
P20_VWRC26217.323_SPEC_EN.pdf P20_VWRC26217.367_SPEC_EN.pdf P20_VWRC26217.460_SPEC_EN.pdf spec_VWRC26217.298 spec_VWRC26217.298
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifikationer
Specifikationstestresultater
- Assay:Min. 98 %
- Refractive index (20°C):1.503 - 1.505
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



