Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Methyl-2-pentanon ≥97%, TECHNICAL

4-Methyl-2-pentanon ≥97%, TECHNICAL

Metal drum, 25 L
Katalog # 25655.466
Brandfarlig
Irriterende
   Sikrere sundhed for mennesker og miljø
Leverandør:   VWR Chemicals
4-Methyl-2-pentanon ≥97%, TECHNICAL
Katalog # 25655.466
Leverandør:  VWR Chemicals
   Sikrere sundhed for mennesker og miljø
CAS nummer:  108-10-1
4-Methyl-2-pentanon ≥97%, TECHNICAL
4-Methyl-2-pentanon ≥97%, TECHNICAL

Metal drum, 25 L

CAS nummer:  108-10-1
Price
6.910,00 kr
Log ind for tilgængelighed
1 * 25 L
Product Details & Documents
Synonymer:  Methylisobutylketon, Isobutylmethylketon
  • CAS nummer:108-10-1
  • Formel:(CH₃)₂CHCH₂COCH₃
  • MDL nummer:MFCD00008938
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,05227
  • Kogende Pt:117,4 °C (1013 hPa)
  • Smeltende Pt:–84 °C
  • Tæthed:0,801 g/cm³ (20 °C)
  • Flash Pt:16 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-550-1
  • FN:1245
  • ADR:3,II
Specifikationer

Specifikationstestresultater

  • Assay:
    Min. 97 %

Specifikationer

  • Pakning:
    Metal drum
  • Pakn:
    25 L
  • Cat. No.:
    25655.466
  • Miljøcertificering:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Sikrere sundhed for mennesker og miljø

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.