Methylethylketon ≥99%, TECHNICAL
Plastic bottle, 1 L
Leverandør: VWR Chemicals
Methylethylketon ≥99%, TECHNICAL
Katalog # 25654.292
Leverandør: VWR Chemicals
CAS nummer: 78-93-3
Gå til:
Product Details & Documents
Synonymer: Ethylmethylketon, Butanon
- CAS nummer:78-93-3
- Formel:C₂H₅COCH₃
- MDL nummer:MFCD00011648
- Molecular Weight:72.11 g/mol
- Merck Index:13,06097
- Kogende Pt:79,6 °C (1013 hPa)
- Smeltende Pt:–86 °C
- Tæthed:0,8054 g/cm³ (20 °C)
- Flash Pt:–4 °C (DIN 51755 Part 1)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-159-0
- FN:1193
- ADR:3,II
- NÅ:01-2119457290-43
Documents
Certifikater
Søg efter partinummer1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Emballageoplysninger
P20_VWRC25654.292_SPEC_EN.pdf P20_VWRC25654.361_SPEC_EN.pdf spec_VWRC25654.292 spec_VWRC25654.320 spec_VWRC25654.361
Produktspecifikation
More
VWR® Green Solvents for Sustainable Chemistry.pdf
Produktlitteratur
Specifikationer
Specifikationstestresultater
- Assay:Min. 99 %
- Boiling point:79 - 80 °C
- Density (20/4):0.804 - 0.807
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 L
- Cat. No.:25654.292
- Miljøcertificering:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



