Methylethylketon ≥99%, GPR RECTAPUR®
Metal drum, 200 L
Leverandør: VWR Chemicals
Methylethylketon ≥99%, GPR RECTAPUR®
Katalog # 25641.550
Leverandør: VWR Chemicals
CAS nummer: 78-93-3
Gå til:
Product Details & Documents
Synonymer: Ethylmethylketon, Butanon
- CAS nummer:78-93-3
- Formel:C₂H₅COCH₃
- MDL nummer:MFCD00011648
- Molecular Weight:72.11 g/mol
- Merck Index:13,06097
- Kogende Pt:79,6 °C (1013 hPa)
- Smeltende Pt:–86 °C
- Tæthed:0,8054 g/cm³ (20 °C)
- Flash Pt:–4 °C (DIN 51755 Part 1)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-159-0
- FN:1193
- ADR:3,II
- NÅ:01-2119457290-43
Certifikater
Søg efter partinummer1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Specifikationer
Specifikationstestresultater
- Assay:Min. 99 %
- IR Spectrum:Passes test
- Boiling point:79 - 80 °C
- Acidity (at filling):Max. 0.0005 meq/g
- Density (20/4):0.804 - 0.807
- Evaporation residue:Max. 20 ppm
- Water:Max. 0.1 %
Specifikationer
- Pakning:Metal drum
- Pakn:200 L
- Cat. No.:25641.550
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



