Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ethylenglycol ≥98%, TECHNICAL

Ethylenglycol ≥98%, TECHNICAL

Plastic bottle, 2,5 L
Katalog # 24407.326
Irriterende
   Sikrere sundhed for mennesker og miljø
Leverandør:   VWR Chemicals
Ethylenglycol ≥98%, TECHNICAL
Katalog # 24407.326
Leverandør:  VWR Chemicals
   Sikrere sundhed for mennesker og miljø
CAS nummer:  107-21-1
Ethylenglycol ≥98%, TECHNICAL
Ethylenglycol ≥98%, TECHNICAL

Plastic bottle, 2,5 L

CAS nummer:  107-21-1
Price
656,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  1,2-Ethandiol, Glykol
  • CAS nummer:107-21-1
  • Formel:HOCH₂CH₂OH
  • MDL nummer:MFCD00002885
  • Molecular Weight:62.07 g/mol
  • Merck Index:13,03832
  • Kogende Pt:196…198 °C (1013 hPa)
  • Smeltende Pt:–13 °C
  • Tæthed:1,115 g/cm³ (20 °C)
  • Flash Pt:116 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-473-3
  • NÅ:01-2119456816-28
Specifikationer

Specifikationstestresultater

  • Assay (on anhydrous substance):
    Min. 98 %
  • Appearance of solution (50 % V/V; water):
    Passes test

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    2,5 L
  • Cat. No.:
    24407.326
  • Miljøcertificering:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Sikrere sundhed for mennesker og miljø

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.