Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D(+)-Glucose monohydrat, powder, TECHNICAL

D(+)-Glucose monohydrat, powder, TECHNICAL

Plastic bucket, 5 kg
Katalog # 24374.366
Leverandør:   VWR Chemicals
D(+)-Glucose monohydrat, powder, TECHNICAL
Katalog # 24374.366
Leverandør:  VWR Chemicals
CAS nummer:  14431-43-7
D(+)-Glucose monohydrat, powder, TECHNICAL
D(+)-Glucose monohydrat, powder, TECHNICAL

Plastic bucket, 5 kg

CAS nummer:  14431-43-7
Price
978,00 kr
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  • CAS Number:14431-43-7
  • Formula:C₆H₁₂O₆·H₂O
  • MDL Number:MFCD00149450
  • Molecular Weight:198.17 g/mol
  • Melting Pt:83 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-075-1
Specifications

Specification Test Results

  • Water:
    7.0 - 9.5 %

Specifications

  • Pakning:
    Plastic bucket
  • Pakn:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.