Distillation apparatus set, general purpose
Product Details & Documents
Fast set-up distillation apparatus for sample pretreatment. Distillation sets include only EPAapproved glassware.
- Easy to set up, only required parts are delivered
- Illustrated, step-by-step procedures simplify distillation
Adapts to a variety of test needs including all wastewater and water methods requiring sample pretreatment by distillation. Applications include fluoride, ammonia-nitrogen, phenols, selenium, volatile acids and albuminoid nitrogen.
Bestillingsoplysninger: Includes distillation flask, J-tube, condenser, drip tube, erlenmeyer flask, and non-Hg thermometer. Arsenic and cyanide sets are available, these are used together with the general purpose distillation set.
Advarsel: Heater and support apparatus, sold separately, is required for distillation.
Specifikationer
- Beskrivelse:Arsenic set