Test tubes with rim
5 ml, 86 mm, 12 mm
Katalog # 212-6721
Leverandør: KIMA SAS DI CHIARIN RENZO
Gå til:
Product Details & Documents
PP, translucent or PS, clear, with rim.
- PS: Good thermal and chemical resistance
- PP: Excellent thermal and chemical resistance, autoclavable
- Robust, economical
Universal tubes for a wide range of applications.
Specifikationer
- Udv.Ø:12 mm
- Kapacitet:5 ml
- Længde:86 mm
- Cat. No.:212-6721