Orders LinkContactUsLinkComponent
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
Løfteplatform till 1× E 106 T/E 206 T/ET 6 and 2× A 24, 110×260 mm
Katalog # 461-2951
Leverandør:   LAUDA
Lifting platforms for Lauda baths
Katalog # 461-2951
Leverandør:  LAUDA
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
Løfteplatform till 1× E 106 T/E 206 T/ET 6 and 2× A 24, 110×260 mm
Price
3.400,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These platforms are made from stainless steel. For use with Lauda baths.
  • Height adjustable in eight steps
  • To be fixed on one side of the bath
  • Not for use with bath cover or gable cover
Specifikationer
  • Beskrivelse:
    Løfteplatform till 1× E 106 T/E 206 T/ET 6 and 2× A 24
  • L×B:
    110×260 mm