Orders LinkContactUsLinkComponent
Lifting platforms for Lauda baths
Løfteplatform til 1× E 4/E 103/E 203; 2× A 12 and 6× A 24, 115×70 mm
Katalog # 461-2948
Leverandør:   LAUDA
Lifting platforms for Lauda baths
Katalog # 461-2948
Leverandør:  LAUDA
Lifting platforms for Lauda baths
Lifting platforms for Lauda baths
Løfteplatform til 1× E 4/E 103/E 203; 2× A 12 and 6× A 24, 115×70 mm
Price
3.340,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These platforms are made from stainless steel. For use with Lauda baths.
  • Height adjustable in eight steps
  • To be fixed on one side of the bath
  • Not for use with bath cover or gable cover
Specifikationer
  • Beskrivelse:
    Løfteplatform til 1× E 4/E 103/E 203; 2× A 12 and 6× A 24
  • L×B:
    115×70 mm
  • Seq:
    01