Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ion selective electrodes, iodide, Orion™

Ion selective electrodes, iodide, Orion™

Iodide solid-state half-cell, 9453BN, 5×10-8 - 1,0 M (5×10-3 - 127000 ppm), 0...+80 °C, 9453BN, BNC
Katalog # 662-0845
Leverandør:   Thermo Orion
Ion selective electrodes, iodide, Orion™
Katalog # 662-0845
Leverandør:  Thermo Orion
Ion selective electrodes, iodide, Orion™

Iodide solid-state half-cell, 9453BN, 5×10-8 - 1,0 M (5×10-3 - 127000 ppm), 0...+80 °C, 9453BN, BNC

Price
8.890,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These electrodes are designed for measuring iodide ions in quickly, simply, accurately and economically.

  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes

Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.

Specifikationer
  • Koncentration:
    5×10-8 - 1,0 M (5×10-3 - 127000 ppm)
  • Tilslutning:
    BNC
  • Model:
    9453BN
  • Beskrivelse:
    Iodide solid-state half-cell, 9453BN
  • Temperatur:
    0...+80 °C
Accessories

Produktinformation

  • Katalog #
    Model
    Pakning
    Tilgængelighed
    Price
  • Ionstyrke justering for Klorid ion selektiv elektrode, i en flaske
    Ionstyrke justering for Klorid ion selektiv elektrode, i en flaske
    662-0824
    Ion selective electrode reagents
    Bottle
    Log ind for tilgængelighed
    1 * 475 mL - 1.180,00 kr

Produktinformation

  • Katalog #
    Koncentration
    Matrice
    Tilgængelighed
    Price
  • Iodid
    Iodid
    662-0844
    0,1M
    H₂O
    Log ind for tilgængelighed
    1 * 475 mL - 1.570,00 kr

Produktinformation

  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Potassium nitrate electrode filling solution for double junction Sure-Flow reference pH electrodes, outer chamber, 60 ml bottle
    Potassium nitrate electrode filling solution for double junction Sure-Flow reference pH electrodes, outer chamber, 60 ml bottle
    662-1524
    5
    Log ind for tilgængelighed
    1 * 5 Items - 1.400,00 kr

Produktinformation

  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Potassium chloride & silver chloride electrode filling solution for double junction Sure-Flow reference pH electrodes, inner chamber, 60 ml bottle
    Potassium chloride & silver chloride electrode filling solution for double junction Sure-Flow reference pH electrodes, inner chamber, 60 ml bottle
    662-0750
    5
    Log ind for tilgængelighed
    1 * 5 Items - 1.400,00 kr