Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,60 ± ml
Katalog # 612-0450
Leverandør:   SciLabware
Volumetric flasks, class A, blue markings
Katalog # 612-0450
Leverandør:  SciLabware
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,60 ± ml
Price
627,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
MBL®, borosilicate glass, with PE stopper.
  • Marks and inscriptions in high contrast blue enamel, conformity 'H' mark
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • Calibrated to contain (TC, In)
 : DIN 12664, BS EN ISO 1042
Specifikationer
  • NS:
    29/32
  • Kapacitet:
    2000 ml
  • Seq:
    11
  • Tolerance:
    0,60 ± ml